COM448622504 Combi-Blocks, Inc.

Methyl 3-(4-nitrophenyl)benzoate

  • MDL number MFCD20529487
  • Cas Number : 107558-26-9
  • Molecular Weight : 257.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-0145

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-0145
Availability details for
Quantity
  • Availability
  • Add to Cart