COM448622528 Combi-Blocks, Inc.

2-[4-(Methylsulfanyl)phenyl]benzonitrile

  • MDL number MFCD05981388
  • Cas Number : 171879-75-7
  • Molecular Weight : 225.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-0182

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-0182
Availability details for
Quantity
  • Availability
  • Add to Cart