COM448622695 Combi-Blocks, Inc.

3-(4-Methylphenyl)benzonitrile

  • MDL number MFCD04038774
  • Cas Number : 133909-96-3
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-0468

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-0468
Availability details for
Quantity
  • Availability
  • Add to Cart