COM448622989 Combi-Blocks, Inc.

N-Cyclopentyl-5-nitropyridin-2-amine

  • MDL number MFCD11120484
  • Cas Number : 939376-51-9
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-0943

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-0943
Availability details for
Quantity
  • Availability
  • Add to Cart