COM448623020 Combi-Blocks, Inc.

2-Methanesulfonylpyridine-4-carbonitrile

  • MDL number MFCD14705772
  • Cas Number : 66154-69-6
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-0978

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?LD-0978
Availability details for
Quantity
  • Availability
  • Add to Cart