COM448623039 Combi-Blocks, Inc.

Methyl 2-methanesulfonylpyridine-3-carboxylate

  • MDL number MFCD23699508
  • Cas Number : 1426958-31-7
  • Molecular Weight : 215.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1009

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1009
Availability details for
Quantity
  • Availability
  • Add to Cart