COM448623067 Combi-Blocks, Inc.

4-Methyl-3-nitrobenzenesulfonohydrazide

  • MDL number MFCD02704624
  • Cas Number : 53516-94-2
  • Molecular Weight : 231.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1057

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1057
Availability details for
Quantity
  • Availability
  • Add to Cart