COM448623096 Combi-Blocks, Inc.

4-Chloro-2-[(1E)-(hydroxyimino)methyl]phenol

  • MDL number MFCD03412413
  • Cas Number : 87974-49-0
  • Molecular Weight : 171.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1103

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1103
Availability details for
Quantity
  • Availability
  • Add to Cart