COM448623116 Combi-Blocks, Inc.

1-(Benzyloxy)-4-ethoxybenzene

  • MDL number MFCD04231644
  • Cas Number : 69697-44-5
  • Molecular Weight : 228.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1130

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1130
Availability details for
Quantity
  • Availability
  • Add to Cart