COM448623118 Combi-Blocks, Inc.

1-(Benzyloxy)-4-propoxybenzene

  • MDL number MFCD28014033
  • Cas Number : 258513-95-0
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1132

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1132
Availability details for
Quantity
  • Availability
  • Add to Cart