COM448623131 Combi-Blocks, Inc.

N-Ethyl-N-(2-nitrophenyl)methanesulfonamide

  • MDL number MFCD23702095
  • Cas Number : 195433-52-4
  • Molecular Weight : 244.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1151

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1151
Availability details for
Quantity
  • Availability
  • Add to Cart