COM448623132 Combi-Blocks, Inc.

N-Isopropyl-N-(2-nitrophenyl)methanesulfonamide

  • MDL number MFCD28096637
  • Cas Number : 195433-54-6
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1152

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1152
Availability details for
Quantity
  • Availability
  • Add to Cart