COM448623136 Combi-Blocks, Inc.

N-Butyl-N-(3-nitrophenyl)methanesulfonamide

  • MDL number MFCD28122684
  • Cas Number : 1820618-28-7
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1156

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1156
Availability details for
Quantity
  • Availability
  • Add to Cart