COM448623138 Combi-Blocks, Inc.

N-Isopropyl-N-(3-nitrophenyl)methanesulfonamide

  • MDL number MFCD28122686
  • Cas Number : 1820641-75-5
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1158

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1158
Availability details for
Quantity
  • Availability
  • Add to Cart