COM448623139 Combi-Blocks, Inc.

N-(2-Nitrophenyl)-N-propylmethanesulfonamide

  • MDL number MFCD28122687
  • Cas Number : 195433-53-5
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1159

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1159
Availability details for
Quantity
  • Availability
  • Add to Cart