COM448623141 Combi-Blocks, Inc.

N-(3-Nitropyridin-2-yl)methanesulfonamide

  • MDL number MFCD28122688
  • Cas Number : 1820716-87-7
  • Molecular Weight : 217.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1167

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1167
Availability details for
Quantity
  • Availability
  • Add to Cart