COM448623142 Combi-Blocks, Inc.

N-(2-Cyanophenyl)-N-isopropylmethanesulfonamide

  • MDL number MFCD28127187
  • Cas Number : 1820735-49-6
  • Molecular Weight : 238.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1168

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1168
Availability details for
Quantity
  • Availability
  • Add to Cart