COM448623154 Combi-Blocks, Inc.

N-(2-Propylaminoethyl)-4-nitroaniline

  • MDL number MFCD16838769
  • Cas Number : 1488659-82-0
  • Molecular Weight : 223.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1185

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1185
Availability details for
Quantity
  • Availability
  • Add to Cart