COM448623159 Combi-Blocks, Inc.

tert-Butyl N-(3-chloro-2-methylphenyl)carbamate

  • MDL number MFCD07127715
  • Cas Number : 129822-39-5
  • Molecular Weight : 241.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1193

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1193
Availability details for
Quantity
  • Availability
  • Add to Cart