COM448623162 Combi-Blocks, Inc.

N-Butyl-N-(4-nitrophenyl)methanesulfonamide

  • MDL number MFCD28252209
  • Cas Number : 1820638-87-6
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1199

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1199
Availability details for
Quantity
  • Availability
  • Add to Cart