COM448623187 Combi-Blocks, Inc.

Lithium triisopropoxy(5-methylthiophen-2-yl)borate

  • MDL number MFCD17015793
  • Cas Number : 1256364-33-6
  • Molecular Weight : 292.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LT-2584

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LT-2584
Availability details for
Quantity
  • Availability
  • Add to Cart