COM448623189 Combi-Blocks, Inc.

Lithium triisopropoxy(4-methoxypyridin-2-yl)boranuide

  • MDL number MFCD28014034
  • Molecular Weight : 303.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LT-4852

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?LT-4852
Availability details for
Quantity
  • Availability
  • Add to Cart