COM448623208 Combi-Blocks, Inc.

Isoquinoline-4-boronic acid, neopentyl glycol ester

  • MDL number MFCD06659058
  • Cas Number : 844891-01-6
  • Molecular Weight : 241.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?NE-2436

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?NE-2436
Availability details for
Quantity
  • Availability
  • Add to Cart