COM448623579 Combi-Blocks, Inc.

2',3',4'-Trihydroxyacetophenone

  • MDL number MFCD00002193
  • Cas Number : 528-21-2
  • Molecular Weight : 168.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-0439

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-0439
Availability details for
Quantity
  • Availability
  • Add to Cart