COM448623587 Combi-Blocks, Inc.

2,4-Dihydroxybenzamide

  • MDL number MFCD00064913
  • Cas Number : 3147-45-3
  • Molecular Weight : 153.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-0447

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-0447
Availability details for
Quantity
  • Availability
  • Add to Cart