COM448623661 Combi-Blocks, Inc.

3-Methyl-2-oxovaleric acid

  • MDL number MFCD00002582
  • Cas Number : 1460-34-0
  • Molecular Weight : 130.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-0535

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-0535
Availability details for
Quantity
  • Availability
  • Add to Cart