COM448623867 Combi-Blocks, Inc.

Synephrine, HCl

  • MDL number MFCD00050566
  • Cas Number : 5985-28-4
  • Molecular Weight : 203.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-0779

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-0779
Availability details for
Quantity
  • Availability
  • Add to Cart