COM448624192 Combi-Blocks, Inc.

8-Azaadenine

  • MDL number MFCD00005697
  • Cas Number : 1123-54-2
  • Molecular Weight : 136.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-1166

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-1166
Availability details for
Quantity
  • Availability
  • Add to Cart