COM448624337 Combi-Blocks, Inc.

4-Butylcyclohexanecarboxylic acid

  • MDL number MFCD00460757
  • Cas Number : 71101-89-8
  • Molecular Weight : 184.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-1337

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-1337
Availability details for
Quantity
  • Availability
  • Add to Cart