COM448624696 Combi-Blocks, Inc.

2,3,5,6-Tetrafluorophenylhydrazine

  • MDL number MFCD00007575
  • Cas Number : 653-11-2
  • Molecular Weight : 180.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-1760

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-1760
Availability details for
Quantity
  • Availability
  • Add to Cart