COM448624785 Combi-Blocks, Inc.

Butyramidine, HCl

  • MDL number MFCD00054341
  • Cas Number : 3020-81-3
  • Molecular Weight : 122.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-1865

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-1865
Availability details for
Quantity
  • Availability
  • Add to Cart