COM448624845 Combi-Blocks, Inc.

(S)-2-tert-Butylcarboxamide-4-tert-butoxycarbonyl piperazine

  • MDL number MFCD02682987
  • Cas Number : 150323-35-6
  • Molecular Weight : 285.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-1938

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-1938
Availability details for
Quantity
  • Availability
  • Add to Cart