COM448624877 Combi-Blocks, Inc.

3-(N-Methylpentylamino)propionic acid, HCl

  • MDL number MFCD08275216
  • Cas Number : 625120-81-2
  • Molecular Weight : 209.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-1978

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-1978
Availability details for
Quantity
  • Availability
  • Add to Cart