COM448624949 Combi-Blocks, Inc.

(R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine

  • MDL number MFCD00010622
  • Cas Number : 72748-99-3
  • Molecular Weight : 130.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-2072

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-2072
Availability details for
Quantity
  • Availability
  • Add to Cart