COM448625339 Combi-Blocks, Inc.

1,4-Diphenyl-1-butanone

  • MDL number MFCD00022005
  • Cas Number : 5407-91-0
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-2616

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-2616
Availability details for
Quantity
  • Availability
  • Add to Cart