COM448625548 Combi-Blocks, Inc.

2-Amino-3,5-dibromo-6-fluorobenzoic acid

  • MDL number MFCD00042156
  • Cas Number : 175135-10-1
  • Molecular Weight : 312.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-2936

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-2936
Availability details for
Quantity
  • Availability
  • Add to Cart