COM448625603 Combi-Blocks, Inc.

3-Cyclopentene-1-carboxylic acid ethyl ester

  • MDL number MFCD06410819
  • Cas Number : 21622-01-5
  • Molecular Weight : 140.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-3001

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-3001
Availability details for
Quantity
  • Availability
  • Add to Cart