COM448625611 Combi-Blocks, Inc.

Morpholin-4-yl-thiophen-2-yl-acetic acid

  • MDL number MFCD06795914
  • Cas Number : 870860-34-7
  • Molecular Weight : 227.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-3011

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-3011
Availability details for
Quantity
  • Availability
  • Add to Cart