COM448625782 Combi-Blocks, Inc.

5-Fluoro-8-hydroxyquinoline

  • MDL number MFCD00059754
  • Cas Number : 387-97-3
  • Molecular Weight : 163.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-3290

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-3290
Availability details for
Quantity
  • Availability
  • Add to Cart