COM448625969 Combi-Blocks, Inc.

(-)-Cis-2-benzamidocyclohexanecarboxylic acid

  • MDL number MFCD00078309
  • Cas Number : 26693-55-0
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-3606

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-3606
Availability details for
Quantity
  • Availability
  • Add to Cart