COM448626009 Combi-Blocks, Inc.

2,3,4-Trifluorocinnamic acid

  • MDL number MFCD00083552
  • Cas Number : 207742-85-6
  • Molecular Weight : 202.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-3667

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-3667
Availability details for
Quantity
  • Availability
  • Add to Cart