COM448626140 Combi-Blocks, Inc.

Bromochlorophen

  • MDL number MFCD00152965
  • Cas Number : 15435-29-7
  • Molecular Weight : 426.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-3900

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-3900
Availability details for
Quantity
  • Availability
  • Add to Cart