COM448626211 Combi-Blocks, Inc.

4-Ethylphenetole

  • MDL number MFCD00191432
  • Cas Number : 1585-06-4
  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4026

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4026
Availability details for
Quantity
  • Availability
  • Add to Cart