COM448626213 Combi-Blocks, Inc.

N-(4-Nitrophenylsulfonyl)-l-phenylalanyl chloride

  • MDL number MFCD00191471
  • Cas Number : 146815-23-8
  • Molecular Weight : 368.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4028

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4028
Availability details for
Quantity
  • Availability
  • Add to Cart