COM448626215 Combi-Blocks, Inc.

5,6,7,8-Tetrahydroquinaldine

  • MDL number MFCD00191581
  • Cas Number : 2617-98-3
  • Molecular Weight : 147.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4031

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4031
Availability details for
Quantity
  • Availability
  • Add to Cart