COM448626270 Combi-Blocks, Inc.

(S)-1-Phenyl-2-(p-tolyl)ethylamine

  • MDL number MFCD00221505
  • Molecular Weight : 211.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4146

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4146
Availability details for
Quantity
  • Availability
  • Add to Cart