COM448626350 Combi-Blocks, Inc.

(R)-(-)-4-Isopropyl-3-propionyl-2-oxazolidinone

  • MDL number MFCD00269691
  • Cas Number : 89028-40-0
  • Molecular Weight : 185.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4274

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4274
Availability details for
Quantity
  • Availability
  • Add to Cart