COM448626384 Combi-Blocks, Inc.

2-Chloro-5-fluoro-3,8-dimethylquinoline

  • MDL number MFCD00276571
  • Cas Number : 175204-94-1
  • Molecular Weight : 209.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4336

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4336
Availability details for
Quantity
  • Availability
  • Add to Cart