COM448626440 Combi-Blocks, Inc.

(R)-2-Isopropylamino-2-phenylethanol

  • MDL number MFCD00671548
  • Cas Number : 112211-92-4
  • Molecular Weight : 179.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4481

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4481
Availability details for
Quantity
  • Availability
  • Add to Cart