COM448626441 Combi-Blocks, Inc.

3-(Methoxycarbonyl)-2h-cyclohepta[b]furan-2-one

  • MDL number MFCD00671552
  • Cas Number : 50603-71-9
  • Molecular Weight : 204.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4482

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4482
Availability details for
Quantity
  • Availability
  • Add to Cart